Learn More
Apexbio Technology LLC LKB1 (AAK1 dual inhibitor) 1093222-27-5 200mg
SDP
Catalog No. 5000064022
Encompass_Preferred
LKB1 (AAK1 dual inhibitor CAS 1093222-27-5) is a selective inhibitor targeting both LKB1 kinase and adaptor-associated kinase 1 (AAK1) These kinases are serine/threonine protein kinases implicated in cellular metabolism polarity and endocytic trafficking pathways respectively By simultaneously inhibiting LKB1 and AAK1 activity this small molecule modulates diverse intracellular signaling cascades associated with cell proliferation and membrane trafficking Due to these mechanistic features LKB1 (AAK1 dual inhibitor) serves as a valuable research tool for studying kinase-associated pathways in various biomedical contexts and potentially informing therapeutic strategies
Catalog No. 50-000-64022 Supplier Apexbio Technology LLC Supplier No. A3556200
LKB1 (AAK1 dual inhibitor CAS 1093222-27-5) is a selective inhibitor targeting both LKB1 kinase and adaptor-associated kinase 1 (AAK1) These kinases are serine/threonine protein kinases implicated in cellular metabolism polarity and endocytic trafficking pathways respectively By simultaneously inhibiting LKB1 and AAK1 activity this small molecule modulates diverse intracellular signaling cascades associated with cell proliferation and membrane trafficking Due to these mechanistic features LKB1 (AAK1 dual inhibitor) serves as a valuable research tool for studying kinase-associated pathways in various biomedical contexts and potentially informing therapeutic strategies