Learn More
Novus Biologicals™ LKB1/STK11 Antibody Blocking Peptide

Specifications
Specifications
| Gene ID (Entrez) | 6794 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-14835. For further blocking peptide related protocol, click here . |
| Gene Alias | EC 2.7.11.1, LKB1 serine/threonine kinase 11 (Peutz-Jeghers syndrome), PJS, PJS polarization-related protein LKB1, Renal carcinoma antigen NY-REN-19, serine/threonine kinase 11, serine/threonine-protein kinase 11, Serine/threonine-protein kinase LKB1, STK11 |
| Gene Symbol | STK11 |
| Quantity | 0.1 mg |
| Research Category | Hypoxia Signaling, mTOR Pathway, p53 Pathway, Phospho Specific, Protein Kinase |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.