Learn More
Eurofins Discoverx LKB1/STRADα/MO25α, active, 10 µg. (14-596)
Supplier:  Eurofins Discoverx 14596
Encompass
N-terminal 6xHis-tagged recombinant, human full length LKB1. Provided as a complex with GST-STRADα and GST-MO25α. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9025