Learn More
Apexbio Technology LLC LL 37(Synonyms: Cathelicidin LL-37, Human Cathelicidin, hCAP-18, CAP18, Antimicrobial peptide LL-37), 1mg, CAS: 154947-66-7.
SDP
Catalog No. 5000061211
Encompass_Preferred
LL-37 (CAS 154947-66-7) is a naturally occurring antimicrobial peptide derived from human cathelicidin consisting of 37 amino acids with amphipathic structure Its mechanism involves interaction with bacterial membranes resulting in membrane disruption and broad-spectrum antibacterial activity In addition to its antimicrobial role LL-37 modulates immune response contributes to inflammatory regulation and participates in wound healing through influencing cell migration and proliferation Current biomedical research employs LL-37 for investigating infection control immune modulation pathways and tissue repair processes
Catalog No. 50-000-61211 Supplier Apexbio Technology LLC Supplier No. B77711
LL-37 (CAS 154947-66-7) is a naturally occurring antimicrobial peptide derived from human cathelicidin consisting of 37 amino acids with amphipathic structure Its mechanism involves interaction with bacterial membranes resulting in membrane disruption and broad-spectrum antibacterial activity In addition to its antimicrobial role LL-37 modulates immune response contributes to inflammatory regulation and participates in wound healing through influencing cell migration and proliferation Current biomedical research employs LL-37 for investigating infection control immune modulation pathways and tissue repair processes