Learn More
Medchemexpress LLC LL-37 amide (TFA) | 99.34% | 4492.28 | 5 MG
SDP
Catalog No. 5000398818
Encompass_Preferred
LL-37 amide TFA is a selective agonist of formyl peptide receptor-like FPRL1, effectively inhibiting periodontal pathogens. It exerts bactericidal effects by activating immune cell chemotaxis and disrupting bacterial cell membrane integrity. This peptide also regulates inflammatory responses and promotes angiogenesis. Its amidation modification reduces sensitivity to serum inhibition and improves stability, making it suitable for research on infection-related diseases like periodontal disease, deep tissue injuries, and wound healing.
  • Selective agonist of formyl peptide receptor-like FPRL1
  • Inhibits periodontal pathogens
  • Exerts bactericidal effects
  • Regulates inflammatory responses
  • Promotes angiogenesis
  • Improved stability and resistance to enzymatic degradation
  • Useful in research for infection-related diseases and wound healing
  • Effective against Gram-positive and Gram-negative bacteria
Catalog No. 50-003-98818 Supplier Medchemexpress LLC Supplier No. HYP4744A5MG
LL-37 amide TFA is a selective agonist of formyl peptide receptor-like FPRL1, effectively inhibiting periodontal pathogens. It exerts bactericidal effects by activating immune cell chemotaxis and disrupting bacterial cell membrane integrity. This peptide also regulates inflammatory responses and promotes angiogenesis. Its amidation modification reduces sensitivity to serum inhibition and improves stability, making it suitable for research on infection-related diseases like periodontal disease, deep tissue injuries, and wound healing.
  • Selective agonist of formyl peptide receptor-like FPRL1
  • Inhibits periodontal pathogens
  • Exerts bactericidal effects
  • Regulates inflammatory responses
  • Promotes angiogenesis
  • Improved stability and resistance to enzymatic degradation
  • Useful in research for infection-related diseases and wound healing
  • Effective against Gram-positive and Gram-negative bacteria