Learn More
Medchemexpress LLC Ll-37 fk-13 tfa | 98.6% | 1719.09 | 5 MG
SDP
Catalog No. 5000396533
Encompass_Preferred
LL-37 FK-13 TFA is the TFA salt form of LL-37 FK-13. It is an antimicrobial agent that inhibits Trichomonas vaginalis. LL-37 FK-13 TFA demonstrates minimal hemolytic effects on human erythrocytes and low cytotoxicity to human fibroblasts.
  • TFA salt form of LL-37 FK-13
  • Antimicrobial agent
  • Inhibits Trichomonas vaginalis
  • Minimal hemolytic effects on human erythrocytes
  • Low cytotoxicity to human fibroblasts
Catalog No. 50-003-96533 Supplier Medchemexpress LLC Supplier No. HYP4836A5MG
LL-37 FK-13 TFA is the TFA salt form of LL-37 FK-13. It is an antimicrobial agent that inhibits Trichomonas vaginalis. LL-37 FK-13 TFA demonstrates minimal hemolytic effects on human erythrocytes and low cytotoxicity to human fibroblasts.
  • TFA salt form of LL-37 FK-13
  • Antimicrobial agent
  • Inhibits Trichomonas vaginalis
  • Minimal hemolytic effects on human erythrocytes
  • Low cytotoxicity to human fibroblasts