Learn More
Apexbio Technology LLC LL-37, human acetate 1mg
SDP
Catalog No. 5000093970
Encompass_Preferred
LL-37 human acetate is an antimicrobial peptide derived from human cathelicidin LL-37 exerts its biological activity primarily by directly interacting with microbial membranes disrupting their integrity and modulating inflammatory signaling pathways which subsequently regulates the secretion of inflammatory mediators Additionally LL-37 influences immune cell activity and cytokine release Based on these pharmacological properties LL-37 human acetate holds research potential in studies of innate immunity host-pathogen interactions inflammatory processes and immunological diseases
Catalog No. 50-000-93970 Supplier Apexbio Technology LLC Supplier No. A90581
LL-37 human acetate is an antimicrobial peptide derived from human cathelicidin LL-37 exerts its biological activity primarily by directly interacting with microbial membranes disrupting their integrity and modulating inflammatory signaling pathways which subsequently regulates the secretion of inflammatory mediators Additionally LL-37 influences immune cell activity and cytokine release Based on these pharmacological properties LL-37 human acetate holds research potential in studies of innate immunity host-pathogen interactions inflammatory processes and immunological diseases