Learn More
Medchemexpress LLC LL-37, human TFA | 154947-66-7 | 100.0% | 4493.26 | 5 MG
SDP
Catalog No. 5000424498
Encompass_Preferred
LL-37, human TFA is a 37-residue, amphipathic, cathelicidin-derived antimicrobial peptide. It demonstrates a broad spectrum of antimicrobial activity.
  • Exhibits broad-spectrum antimicrobial activity.
  • May help protect the cornea from infection.
  • Modulates wound healing.
Catalog No. 50-004-24498 Supplier Medchemexpress LLC Supplier No. HYP1222A5MG
LL-37, human TFA is a 37-residue, amphipathic, cathelicidin-derived antimicrobial peptide. It demonstrates a broad spectrum of antimicrobial activity.
  • Exhibits broad-spectrum antimicrobial activity.
  • May help protect the cornea from infection.
  • Modulates wound healing.