Learn More
AURICAL LLC LL CNDCTVTY STD 2.0US/CM 500ML
Catalog No. NC9981948
Encompass_Preferred
Low Level Conductivity Standard 2.0�s/cm 500 mL pouches for accurate conductivity measurement in purified water. Standard for accurate conductivity measurement in purified water. High accuracy is required for pure water and water for injection in pharmaceutical manufacturing.
Catalog No. NC9981948 Supplier AURICAL LLC Supplier No. 300022
Low Level Conductivity Standard 2.0�s/cm 500 mL pouches for accurate conductivity measurement in purified water. Standard for accurate conductivity measurement in purified water. High accuracy is required for pure water and water for injection in pharmaceutical manufacturing.