Learn More
Metrohm USA LL ISE REFERENCE ELECTRODE
SDP
Catalog No. NC0243487 Shop All Metrohm USA Products
Encompass
Double-junction Ag/AgCl reference electrode with fixed ground-joint diaphragm, minimum immersion depth 1 mm.
Catalog No. NC0243487 Supplier Metrohm USA Supplier No. 60750100
Double-junction Ag/AgCl reference electrode with fixed ground-joint diaphragm, minimum immersion depth 1 mm.