Learn More
Cayman Chemical LL-Z 1640-4, Purity: ≥95%, CAS: 66018-41-5, 5 mg
SDP
Catalog No. 502985182
Encompass_Preferred
A resorcylic acid lactone with antiviral and antiprotozoan activity; used as negative control for MAP3K inhibitor (5Z)-7-oxo zeaenol (Item No. 17459)
Catalog No. 50-298-5182 Supplier Cayman Chemical Supplier No. 191595 MG
A resorcylic acid lactone with antiviral and antiprotozoan activity; used as negative control for MAP3K inhibitor (5Z)-7-oxo zeaenol (Item No. 17459)