Learn More
ATCC LL/2 (LLC1); Lewis Lung Carcinoma; Mouse (Mus musculus)
Catalog No. 502383113
Encompass
LL/2 (LLC1) Lewis lung carcinoma is a cell line established from the lung of a C57BL mouse bearing a tumor resulting from an implantation of primary Lewis lung carcinoma. This cell line is widely used as a model for metastasis and is useful for studying the mechanisms of cancer chemotherapeutic agents.
Catalog No. 50-238-3113 Supplier ATCC Supplier No. CRL1642
 Restricted Item
Add to Cart
Add to Cart
LL/2 (LLC1) Lewis lung carcinoma is a cell line established from the lung of a C57BL mouse bearing a tumor resulting from an implantation of primary Lewis lung carcinoma. This cell line is widely used as a model for metastasis and is useful for studying the mechanisms of cancer chemotherapeutic agents.