Learn More
Cayman Chemical L-Leucine 4-methoxy-β-naphthylamide (hydrochloride), Purity: ≥98%, CAS: 4467-68-9, 100 mg
SDP
Catalog No. 502968970
Encompass_Preferred
A cell-permeable substrate for aminopeptidase M and leucine aminopeptidase that was developed for intracellular analysis of protease activities
Catalog No. 50-296-8970 Supplier Cayman Chemical Supplier No. 16223100 MG
A cell-permeable substrate for aminopeptidase M and leucine aminopeptidase that was developed for intracellular analysis of protease activities