Learn More
Medchemexpress LLC LLO (91-99) | 137856-41-8 | 99.2% | 1058.10 | 1 MG
SDP
Catalog No. 5000423597
Encompass_Preferred
LLO (91-99) is an exotoxin and a class I MHC-restricted T-cell epitope of listeriolysin (LLO). It is processed by antigen-presenting cells, binds to MHC class I molecules, and is presented to the cell surface. It induces CTL responses and effectively eliminates Listeria from infected cells. It can be used in the research of Listeria infections (e.g., listeriosis).
  • Induces cytotoxic T lymphocyte (CTL) responses
  • Effectively eliminates Listeria from infected cells
  • Suitable for research on Listeria infections like listeriosis
  • Pulsed dendritic cells show high protection efficiency against listeriosis
  • Induces strong CD8+ T cell responses with IFN-γ production
  • Loaded DCs protect C57BL/6 mice against Listeria monocytogenes infection
  • Reduces liver granulomatous lesions
Catalog No. 50-004-23597 Supplier Medchemexpress LLC Supplier No. HYP24551MG
Add to Cart
Add to Cart
LLO (91-99) is an exotoxin and a class I MHC-restricted T-cell epitope of listeriolysin (LLO). It is processed by antigen-presenting cells, binds to MHC class I molecules, and is presented to the cell surface. It induces CTL responses and effectively eliminates Listeria from infected cells. It can be used in the research of Listeria infections (e.g., listeriosis).
  • Induces cytotoxic T lymphocyte (CTL) responses
  • Effectively eliminates Listeria from infected cells
  • Suitable for research on Listeria infections like listeriosis
  • Pulsed dendritic cells show high protection efficiency against listeriosis
  • Induces strong CD8+ T cell responses with IFN-γ production
  • Loaded DCs protect C57BL/6 mice against Listeria monocytogenes infection
  • Reduces liver granulomatous lesions