Learn More
PLANT CELL TECHNOLOGY Lloyd & McCown's Woody Plant Basal Medium With Macronutrients, Micronutrients and Vitamins, Plant Tissue Culture, Mod. 4, Off-White to Yellow Powder, 50L Bottle
Catalog No. NC3905213
Encompass_Preferred
Woody Plant Basal Medium Mod 4 is a complete formulation for micropropagation of woody plant species. Supplied in a screw-top bottle, this 50L equivalent of off white to yellow powder is ideal for use in research labs, commercial propagation, and educational applications. Stable under recommended storage conditions, ensuring reproducible tissue culture results.
Catalog No. NC3905213 Supplier PLANT CELL TECHNOLOGY Supplier No. WPP0450LT
Woody Plant Basal Medium Mod 4 is a complete formulation for micropropagation of woody plant species. Supplied in a screw-top bottle, this 50L equivalent of off white to yellow powder is ideal for use in research labs, commercial propagation, and educational applications. Stable under recommended storage conditions, ensuring reproducible tissue culture results.