Learn More
Medchemexpress LLC LLVK TFA | 585.66 | 25 MG
SDP
Catalog No. 5000426998
Encompass_Preferred
LLVK TFA is a selective IκB phosphorylation inhibitor. It reduces LPS-induced pro-inflammatory cytokines (IL-1β, IL-6, TNF-α). LLVK TFA is promising for research of inflammatory bowel disease and rheumatoid arthritis.
  • Selective IκB phosphorylation inhibitor
  • Reduces LPS-induced pro-inflammatory cytokines (IL-1β, IL-6, TNF-α)
  • Promising for research of inflammatory bowel disease
  • Promising for research of rheumatoid arthritis
Catalog No. 50-004-26998 Supplier Medchemexpress LLC Supplier No. 25MGHYP11296A
LLVK TFA is a selective IκB phosphorylation inhibitor. It reduces LPS-induced pro-inflammatory cytokines (IL-1β, IL-6, TNF-α). LLVK TFA is promising for research of inflammatory bowel disease and rheumatoid arthritis.
  • Selective IκB phosphorylation inhibitor
  • Reduces LPS-induced pro-inflammatory cytokines (IL-1β, IL-6, TNF-α)
  • Promising for research of inflammatory bowel disease
  • Promising for research of rheumatoid arthritis