Learn More
TARGETMOL CHEMICALS INC LLY-283 5MG
SDP
Catalog No. 502581204
Encompass_Preferred
Also available in 1 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LLY-283 is a potent selective and oral protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 22 nM and a Kd of 6 nM for PRMT5MEP50 complex and shows antitumor activity. purity: 98%
Catalog No. 50-258-1204 Supplier TARGETMOL CHEMICALS INC Supplier No. T157675MG
Also available in 1 mg 10 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LLY-283 is a potent selective and oral protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 22 nM and a Kd of 6 nM for PRMT5MEP50 complex and shows antitumor activity. purity: 98%