Learn More
Medchemexpress LLC HY-107777 5mg Medchemexpress, LLY-283 CAS:2040291-27-6 Purity:>98%
SDP
Catalog No. 501932296
Encompass_Preferred
Medchemexpress, HY-107777 5mg LLY-283 CAS:2040291-27-6 LLY-283 is a potent, selective and oral protein arginine methyltransferase 5 ( PRMT5 ) inhibitor, with an IC 50 of 22 nM and a K d of 6 nM for PRMT5:MEP50 complex, and shows antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2296 Supplier Medchemexpress LLC Supplier No. HY1077775MG
Medchemexpress, HY-107777 5mg LLY-283 CAS:2040291-27-6 LLY-283 is a potent, selective and oral protein arginine methyltransferase 5 ( PRMT5 ) inhibitor, with an IC 50 of 22 nM and a K d of 6 nM for PRMT5:MEP50 complex, and shows antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.