Learn More
Medchemexpress LLC LLY-507 | 1793053-37-8 | 99.09% | 574.76 | 50 MG
SDP
Catalog No. 5000385868
Encompass_Preferred
LLY-507 is a potent and selective inhibitor of protein-lysine methyltransferase SMYD2. It potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM. LLY-507 serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes. It is for research use only.
  • Potent and selective inhibitor of protein-lysine methyltransferase SMYD2.
  • Potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM.
  • Serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes.
Catalog No. 50-003-85868 Supplier Medchemexpress LLC Supplier No. HY1931350MG
LLY-507 is a potent and selective inhibitor of protein-lysine methyltransferase SMYD2. It potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM. LLY-507 serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes. It is for research use only.
  • Potent and selective inhibitor of protein-lysine methyltransferase SMYD2.
  • Potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM.
  • Serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes.