Learn More
Medchemexpress LLC LLY-507 | 1793053-37-8 | 99.1% | 574.76 | 25 MG
SDP
Catalog No. 5000385908
Encompass_Preferred
LLY-507 is a potent and selective inhibitor of protein-lysine methyltransferase SMYD2. It potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM. This compound serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes.
  • Potent and selective inhibitor of SMYD2.
  • Inhibits SMYD2 from methylating p53 peptide with an IC50 <15 nM.
  • Useful chemical probe for dissecting SMYD2 function in cancer and other biological processes.
Catalog No. 50-003-85908 Supplier Medchemexpress LLC Supplier No. 25MGHY19313
LLY-507 is a potent and selective inhibitor of protein-lysine methyltransferase SMYD2. It potently inhibits the ability of SMYD2 to methylate p53 peptide with an IC50 <15 nM. This compound serves as a valuable chemical probe to aid in the dissection of SMYD2 function in cancer and other biological processes.
  • Potent and selective inhibitor of SMYD2.
  • Inhibits SMYD2 from methylating p53 peptide with an IC50 <15 nM.
  • Useful chemical probe for dissecting SMYD2 function in cancer and other biological processes.