Learn More
Selleck Chemical LLC LLY-507
SDP
Catalog No. 5000004461
Encompass_Preferred
LLY-507 is a cell-active potent and selective inhibitor of protein-lysine Methyltransferase SMYD2
Catalog No. 50-000-04461 Supplier Selleck Chemical LLC Supplier No. S757525MG
Add to Cart
Add to Cart
LLY-507 is a cell-active potent and selective inhibitor of protein-lysine Methyltransferase SMYD2