Learn More
Cayman Chemical LLysIn lactamhydrochLRId 50mg
SDP
Catalog No. 502991043
Encompass_Preferred
A building block; has been used in the synthesis of HIV protease inhibitors, as well as bengamide derivatives with in vitro anticancer activity
Catalog No. 50-299-1043 Supplier Cayman Chemical Supplier No. 3168550 MG
A building block; has been used in the synthesis of HIV protease inhibitors, as well as bengamide derivatives with in vitro anticancer activity