Learn More
Cayman Chemical L-Lysine lactam (hydrochloride), Purity: ≥95%, CAS: 26081-07-2, 100 mg
SDP
Catalog No. 502978969
Encompass_Preferred
A building block; has been used in the synthesis of HIV protease inhibitors, as well as bengamide derivatives with in vitro anticancer activity
Catalog No. 50-297-8969 Supplier Cayman Chemical Supplier No. 31685100 MG
A building block; has been used in the synthesis of HIV protease inhibitors, as well as bengamide derivatives with in vitro anticancer activity