Learn More
Electron Microscopy Sciences Light Microscopy Perfect Loop Set
SDP
Catalog No. 50930554
Encompass_Preferred
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases. For light microscopy (large sections). Outside diameter of loop is 7mm. Set includes loop and handle.
Catalog No. 50-930-554 Supplier Electron Microscopy Sciences Supplier No. 70940
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases. For light microscopy (large sections). Outside diameter of loop is 7mm. Set includes loop and handle.