Learn More
Motic™ LM Plan Achromatic Objective
SCI_ED
Catalog No. 01674235 Shop All Motic Group Products
Change view
Click to view available options
Magnification Power:
100X
Catalog No. Magnification Power
01-674-235 100X
1 options
This item is not returnable. View return policy
Catalog No. 01-674-235 Supplier Motic™ Supplier No. 1101001704512
Add to Cart
Add to Cart
This item is not returnable. View return policy
The BA310MET series presents an affordable and powerful microscope line for incident light applications in all material sciences.
  • Objectives with long working distance for incident light provide optimal image contrast for reflective, non-covered specimen
  • Together with a fully corrected tube lens, an intermediate image without colored fringes is created
  • 0.8 N.A.
Specifications
Type LM Plan Achromatic Objective
For Use With (Equipment) BA310MET Series
Magnification Power 100X
Working Distance 2 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov