Learn More
DYNAMIC DEVICES LM Series, 1250 μl Disp. Tips, Sterile & FilteredCase of DDX Series 1250μl disposable tips
Catalog No. NC1825010
Encompass_Preferred
This item has a minimum order quantity of 5 cases per supplier requirements.
LM Series, 1250 μl Disp. Tips, Sterile & FilteredCase of DDX Series 1250μl disposable tips, Sterile & Filtered.For the Lynx DDX Series 96 Tip Pipette Head. Each Caseincludes 3,840 total tips packaged in 40 racks of 96 tips in arobotic box.
Catalog No. NC1825010 Supplier DYNAMIC DEVICES Supplier No. DDX961250F5
This item has a minimum order quantity of 5 cases per supplier requirements.
LM Series, 1250 μl Disp. Tips, Sterile & FilteredCase of DDX Series 1250μl disposable tips, Sterile & Filtered.For the Lynx DDX Series 96 Tip Pipette Head. Each Caseincludes 3,840 total tips packaged in 40 racks of 96 tips in arobotic box.