Learn More
DYNAMIC DEVICES DDX 300ul Tips, Non-Filtered, 4,800 total tips packaged in 50 racks of 96 tips per case
Catalog No. NC2820362
Encompass
Pipette Head: 96VVP, 96ST, f8VVP, f4iZ VVP
• 300ul
• Non-Filtered
• Each Case includes 4,800 total tips packaged in 50 racks of 96 tips in a robotic box
Catalog No. NC2820362 Supplier DYNAMIC DEVICES Supplier No. DDX96300NF1
Pipette Head: 96VVP, 96ST, f8VVP, f4iZ VVP
• 300ul
• Non-Filtered
• Each Case includes 4,800 total tips packaged in 50 racks of 96 tips in a robotic box