Learn More
Alkali Scientific LM Sieve Agarose, 500 G
SDP
Catalog No. 5000661107
Encompass_Preferred
Low Melting Sieve Agarose is a specialized agarose designed for DNA gel electrophoresis. It has a lower melting point compared to standard agarose, making it ideal for the separation of DNA fragments at lower temperatures. This 500g package is used for preparing gels that are ideal for applications requiring low melting points, such as DNA fragment separation, electrophoresis, and enzymatic reactions. The agarose is pre-formulated for easy use in laboratories that routinely conduct gel electrophoresis for DNA analysis. It offers reliable resolution, sharp band separation, and high sensitivity, ensuring reproducibility in molecular biology and genetic research applications.
Catalog No. 50-006-61107 Supplier Alkali Scientific Supplier No. LMS002CP
Low Melting Sieve Agarose is a specialized agarose designed for DNA gel electrophoresis. It has a lower melting point compared to standard agarose, making it ideal for the separation of DNA fragments at lower temperatures. This 500g package is used for preparing gels that are ideal for applications requiring low melting points, such as DNA fragment separation, electrophoresis, and enzymatic reactions. The agarose is pre-formulated for easy use in laboratories that routinely conduct gel electrophoresis for DNA analysis. It offers reliable resolution, sharp band separation, and high sensitivity, ensuring reproducibility in molecular biology and genetic research applications.