Learn More
Medchemexpress LLC LM10 | 1316695-35-8 | 99.9% | 229.22 | 100 MG
SDP
Catalog No. 5000403766
Encompass_Preferred
LM10 is a potent inhibitor of tryptophan 2,3-dioxygenase (TDO). Tryptophan 2,3-dioxygenase (TDO) is an unrelated hepatic enzyme that also degrades tryptophan along the kynurenine pathway. LM10 has the potential for the research of cancer diseases.
  • Prevents the growth of TDO-expressing P815 tumor cells and promotes better rejection of control clone P815B cl1, which does not express TDO.
  • Displays a good TDO inhibition (Ki = 5.6 μM) with a competitive inhibition profile.
  • Does not inhibit IDO and has a high solubility and bioavailability without obvious signs of toxicity.
  • The plasma concentration after oral administration is about 40 times above the IC50 measured in the cellular assay performed with the physiological concentration of plasma tryptophan.
Catalog No. 50-004-03766 Supplier Medchemexpress LLC Supplier No. HY33298100MG
LM10 is a potent inhibitor of tryptophan 2,3-dioxygenase (TDO). Tryptophan 2,3-dioxygenase (TDO) is an unrelated hepatic enzyme that also degrades tryptophan along the kynurenine pathway. LM10 has the potential for the research of cancer diseases.
  • Prevents the growth of TDO-expressing P815 tumor cells and promotes better rejection of control clone P815B cl1, which does not express TDO.
  • Displays a good TDO inhibition (Ki = 5.6 μM) with a competitive inhibition profile.
  • Does not inhibit IDO and has a high solubility and bioavailability without obvious signs of toxicity.
  • The plasma concentration after oral administration is about 40 times above the IC50 measured in the cellular assay performed with the physiological concentration of plasma tryptophan.