Learn More
Medchemexpress LLC Lm10 | 1316695-35-8 | 99.9% | 229.22 | 50 MG
SDP
Catalog No. 5000404344
Encompass_Preferred
LM10 is a potent inhibitor of tryptophan 2,3-dioxygenase (TDO), an hepatic enzyme that degrades tryptophan along the kynurenine pathway. This compound shows potential for research in cancer diseases. It exhibits good TDO inhibition (Ki = 5.6 μM) with a competitive inhibition profile and does not inhibit IDO. LM10 has high solubility and bioavailability without obvious signs of toxicity.
  • Potent inhibitor of tryptophan 2,3-dioxygenase (TDO).
  • Potential for cancer disease research.
  • Displays good TDO inhibition (Ki = 5.6 μM) with a competitive inhibition profile.
  • Does not inhibit IDO.
  • High solubility and bioavailability.
  • No obvious signs of toxicity.
  • Prevents the growth of TDO-expressing P815 tumor cells in vivo.
  • Promotes better rejection of control clone P815B cl1, which does not express TDO, in vivo.
Catalog No. 50-004-04344 Supplier Medchemexpress LLC Supplier No. HY3329850MG
LM10 is a potent inhibitor of tryptophan 2,3-dioxygenase (TDO), an hepatic enzyme that degrades tryptophan along the kynurenine pathway. This compound shows potential for research in cancer diseases. It exhibits good TDO inhibition (Ki = 5.6 μM) with a competitive inhibition profile and does not inhibit IDO. LM10 has high solubility and bioavailability without obvious signs of toxicity.
  • Potent inhibitor of tryptophan 2,3-dioxygenase (TDO).
  • Potential for cancer disease research.
  • Displays good TDO inhibition (Ki = 5.6 μM) with a competitive inhibition profile.
  • Does not inhibit IDO.
  • High solubility and bioavailability.
  • No obvious signs of toxicity.
  • Prevents the growth of TDO-expressing P815 tumor cells in vivo.
  • Promotes better rejection of control clone P815B cl1, which does not express TDO, in vivo.