Learn More
Medchemexpress LLC LM11A-31 10mM 1mL | 102562-74-3 | 243.35 g/mol | C12H25N3O2 | 1 ML
SDP
Catalog No. 5000229243
Encompass_Preferred
LM11A-31 a non-peptide p75NTR (neurotrophin receptor p75) modulator is an orally active and potent proNGF (nerve growth factor) antagonist LM11A-31 is an amino acid derivative with high blood-brain barrier permeability and blocks p75-mediated cell death LM11A-31 reverses cholinergic neurite dystrophy in Alzheimer s disease mouse models with mid- to late-stage disease progression[1][2]
Catalog No. 50-002-29243 Supplier Medchemexpress LLC Supplier No. HY11708810MM 1ML
LM11A-31 a non-peptide p75NTR (neurotrophin receptor p75) modulator is an orally active and potent proNGF (nerve growth factor) antagonist LM11A-31 is an amino acid derivative with high blood-brain barrier permeability and blocks p75-mediated cell death LM11A-31 reverses cholinergic neurite dystrophy in Alzheimer s disease mouse models with mid- to late-stage disease progression[1][2]