Learn More
Medchemexpress LLC LM11A-31 5mg | 102562-74-3 | 243.35 g/mol | C12H25N3O2 | 5 MG
SDP
Catalog No. 5000229077
Encompass_Preferred
LM11A-31 is a small-molecule ligand and modulator of the p75 neurotrophin receptor (p75NTR) used in preclinical research of neurodegenerative diseases. It is orally active, brain-penetrant, and provided for in vitro and in vivo studies in solid and solution formats.
  • Small-molecule p75NTR modulator for neurodegeneration research.
  • Orally active and brain-penetrant in preclinical studies.
  • Available as a solid and as 10 mM solutions in DMSO for convenience.
  • Identified by CAS 102562-74-3 and molecular formula C12H25N3O2.
  • High reported purity (~99.9%) suitable for research applications.
  • Commonly supplied in small research pack sizes such as 5 mg.
Catalog No. 50-002-29077 Supplier Medchemexpress LLC Supplier No. HY1170885MG
LM11A-31 is a small-molecule ligand and modulator of the p75 neurotrophin receptor (p75NTR) used in preclinical research of neurodegenerative diseases. It is orally active, brain-penetrant, and provided for in vitro and in vivo studies in solid and solution formats.
  • Small-molecule p75NTR modulator for neurodegeneration research.
  • Orally active and brain-penetrant in preclinical studies.
  • Available as a solid and as 10 mM solutions in DMSO for convenience.
  • Identified by CAS 102562-74-3 and molecular formula C12H25N3O2.
  • High reported purity (~99.9%) suitable for research applications.
  • Commonly supplied in small research pack sizes such as 5 mg.