Learn More
Medchemexpress LLC LM11A-31 dihydrochloride | 1243259-19-9 | 99.9% | 316.27 | 10 MG
SDP
Catalog No. 5000343310
Encompass_Preferred
LM11A-31 dihydrochloride is a non-peptide p75NTR (neurotrophin receptor p75) modulator and an orally active, potent proNGF (nerve growth factor) antagonist. It is an amino acid derivative with high blood-brain barrier permeability that blocks p75-mediated cell death. This compound reverses cholinergic neurite dystrophy in Alzheimer's disease mouse models at mid- to late-stage disease progression. It is for research use only.
  • Non-peptide p75NTR modulator
  • Orally active and potent proNGF antagonist
  • Amino acid derivative
  • High blood-brain barrier permeability
  • Blocks p75-mediated cell death
  • Reverses cholinergic neurite dystrophy in Alzheimer's disease mouse models
  • Mitigates proNGF accumulation and preserves BRB integrity (in vivo)
  • Prevents and/or reverses atrophy of basal forebrain cholinergic neurites and cortical dystrophic neurites in mid-stage male APPL/S mice (in vivo)
Catalog No. 50-003-43310 Supplier Medchemexpress LLC Supplier No. HY11015510MG
LM11A-31 dihydrochloride is a non-peptide p75NTR (neurotrophin receptor p75) modulator and an orally active, potent proNGF (nerve growth factor) antagonist. It is an amino acid derivative with high blood-brain barrier permeability that blocks p75-mediated cell death. This compound reverses cholinergic neurite dystrophy in Alzheimer's disease mouse models at mid- to late-stage disease progression. It is for research use only.
  • Non-peptide p75NTR modulator
  • Orally active and potent proNGF antagonist
  • Amino acid derivative
  • High blood-brain barrier permeability
  • Blocks p75-mediated cell death
  • Reverses cholinergic neurite dystrophy in Alzheimer's disease mouse models
  • Mitigates proNGF accumulation and preserves BRB integrity (in vivo)
  • Prevents and/or reverses atrophy of basal forebrain cholinergic neurites and cortical dystrophic neurites in mid-stage male APPL/S mice (in vivo)