Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lm11a-31 dihydrochloride | 1243259-19-9 | MFCD13245016 | 99.9% | 316.27 g/mol | C12H27Cl2N3O2 | 5 MG
SDP

Catalog No. 5000263297
Encompass_Preferred

LM11A-31 dihydrochloride is a brain-penetrant, small-molecule modulator of the p75 neurotrophin receptor that acts as a potent proNGF antagonist. It is supplied for research use and has been reported to reverse cholinergic neurite dystrophy in Alzheimer's disease models.

  • Brain-penetrant small molecule p75NTR modulator and proNGF antagonist.
  • Reported to reverse cholinergic neurite dystrophy in Alzheimer's disease models.
  • High purity (99.93%).
  • Soluble in DMSO and water at up to 100 mg/mL; compatible with PBS for in vivo formulations.
  • Available in small research sizes (for example, 5 mg) and larger quantities on request.
  • Recommended storage: 4°C short-term; in solvent, -80°C up to 6 months and -20°C up to 1 month under inert atmosphere.

Catalog No. 50-002-63297 Supplier Medchemexpress LLC Supplier No. HY1101555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LM11A-31 dihydrochloride is a brain-penetrant, small-molecule modulator of the p75 neurotrophin receptor that acts as a potent proNGF antagonist. It is supplied for research use and has been reported to reverse cholinergic neurite dystrophy in Alzheimer's disease models.

  • Brain-penetrant small molecule p75NTR modulator and proNGF antagonist.
  • Reported to reverse cholinergic neurite dystrophy in Alzheimer's disease models.
  • High purity (99.93%).
  • Soluble in DMSO and water at up to 100 mg/mL; compatible with PBS for in vivo formulations.
  • Available in small research sizes (for example, 5 mg) and larger quantities on request.
  • Recommended storage: 4°C short-term; in solvent, -80°C up to 6 months and -20°C up to 1 month under inert atmosphere.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.