Learn More
Cayman Chemical LM11A-31 (hydrochloride), Purity: ≥95%, CAS: 1243259-19-9, 50 mg
SDP
Catalog No. 502987405
Encompass_Preferred
A p75NTR ligand; inhibits NGF binding to p75NTR-Fc (A2 = 1,192 nM); inhibits DNA damage in and promotes survival of hippocampal neuronal cultures in a p75NTR- and concentration-dependent manner; prevents neurodegeneration in various mouse models of injury and neurodegenerative disease
Catalog No. 50-298-7405 Supplier Cayman Chemical Supplier No. 2198250 MG
A p75NTR ligand; inhibits NGF binding to p75NTR-Fc (A2 = 1,192 nM); inhibits DNA damage in and promotes survival of hippocampal neuronal cultures in a p75NTR- and concentration-dependent manner; prevents neurodegeneration in various mouse models of injury and neurodegenerative disease