Learn More
Medchemexpress LLC HY-100673 5mg Medchemexpress, LM22A-4 CAS:37988-18-4 Purity:>98%
SDP
Catalog No. 501931913
Encompass_Preferred
Medchemexpress, HY-100673 5mg LM22A-4 CAS:37988-18-4 LM22A-4 is a specific agonist of tyrosine kinase receptor B , used for neurological disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1913 Supplier Medchemexpress LLC Supplier No. HY1006735MG
Medchemexpress, HY-100673 5mg LM22A-4 CAS:37988-18-4 LM22A-4 is a specific agonist of tyrosine kinase receptor B , used for neurological disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.