Learn More
Abcam LM22A-4, neurotrophic BDNF mimetic, 50MG
Catalog No. 502702912
Encompass_Preferred
MW 339.34 Da, Purity >98%. Small molecule BDNF mimetic displaying neurotrophic activity. Partial agonist at TrkB, activating TrkB signaling. Displaces BDNF (IC₅₀ = 47 nM) in binding assays. Prevents neuronal degeneration with equal efficacy to that of BDNF in in vitro models of neurodegenerative disease. Causes hippocampal and striatal TrkB activation in mice and improved motor learning after traumatic brain injury in rats, after in vivo administraton. Improves respiratory function in a mouse model of Rett syndrome.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-2912 Supplier Abcam Supplier No. AB22202050MG
MW 339.34 Da, Purity >98%. Small molecule BDNF mimetic displaying neurotrophic activity. Partial agonist at TrkB, activating TrkB signaling. Displaces BDNF (IC₅₀ = 47 nM) in binding assays. Prevents neuronal degeneration with equal efficacy to that of BDNF in in vitro models of neurodegenerative disease. Causes hippocampal and striatal TrkB activation in mice and improved motor learning after traumatic brain injury in rats, after in vivo administraton. Improves respiratory function in a mouse model of Rett syndrome.
The product is subject to the following: Abcam Restricted Use Statement