Learn More
TARGETMOL CHEMICALS INC LM22B-10 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T44965MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LM22B-10 is an activator of TrkB/TrkC neurotrophin receptor and can induce TrkB TrkC ERK and AKT activation in vitro and in vivo. purity: 98%
Catalog No. 50-257-6758