Learn More
Aviva Systems Biology Corp LMAN1 Antibody - N-terminal region (ARP46613_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46613_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LMAN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LMAN1. Alternate names: ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59361