Learn More
Aviva Systems Biology Corp LMAN2 Antibody - N-terminal region (ARP46788_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46788_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LMAN2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human LMAN2. Alternate names: C5orf8, GP36B, VIP36ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59398