Learn More
ABclonal Technology LMAN2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A153941000UL
Encompass_Preferred
This gene encodes a type I transmembrane lectin that shuttles between the endoplasmic reticulum, the Golgi apparatus and the plasma membrane. The encoded protein binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control.
Catalog No. 50-001-06070