Learn More
Aviva Systems Biology Corp LMBR1 Antibody - N-terminal region (ARP49722_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49722_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LMBR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LMBR1. Alternate names: ACHP, C7orf2, DIF14, FLJ11665, PPD2, TPTApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60159