Learn More
Selleck Chemical LLC LMK-235
SDP
Supplier:  Selleck Chemical LLC S75691G
Encompass_Preferred
LMK-235 is a selective inhibitor of HDAC4 and HDAC5 with IC50 of 11 9 nM and 4 2 nM respectively
Catalog No. 50-000-04696