Learn More
Abcam LMK 235, HDAC inhibitor, 5MG
Catalog No. 502672393
Encompass_Preferred
MW 294.35 Da, Purity >98%. Highly potent and selective HDAC inhibitor. Greater potency for HDAC4 and HDAC5 (IC₅₀ = 11.9 and 4.2 nM, respectively) than other HDAC family members (IC₅₀ values = HDAC1 320 nM, HDAC2 881 nM, HDAC6 55.7 nM, and HDAC8 1278 nM. Demonstrates inhibitor activity in human ovarian cell lines (IC₅₀ =0.49μM) and cisplatin resistant cell lines (IC₅₀ = 0.32μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2393 Supplier Abcam Supplier No. AB2543285MG
MW 294.35 Da, Purity >98%. Highly potent and selective HDAC inhibitor. Greater potency for HDAC4 and HDAC5 (IC₅₀ = 11.9 and 4.2 nM, respectively) than other HDAC family members (IC₅₀ values = HDAC1 320 nM, HDAC2 881 nM, HDAC6 55.7 nM, and HDAC8 1278 nM. Demonstrates inhibitor activity in human ovarian cell lines (IC₅₀ =0.49μM) and cisplatin resistant cell lines (IC₅₀ = 0.32μM).
The product is subject to the following: Abcam Restricted Use Statement