Learn More
Aviva Systems Biology Corp Lmo2 Antibody - C-terminal region (ARP38632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lmo2. It was validated on Western Blot. . Alternate names: Rbtn-2, Rbtn2, Rhom-2, Ttg2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-748