Learn More
Aviva Systems Biology Corp Lmo3 Antibody - N-terminal region (ARP38139_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38139_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lmo3. It was validated on Western Blot. . Alternate names: AI854781, BB106490, Rbtn-3, Rbtn3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-722