Learn More
Aviva Systems Biology Corp LMOD1 Antibody - N-terminal region (ARP54847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LMOD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LMOD1. Alternate names: 1D, 64kD, D1, SM-LMODApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61341