Learn More
Aviva Systems Biology Corp Lmod1 Antibody - C-terminal region (ARP54848_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54848_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Lmod1. It was validated on Western Blot. . Alternate names: 9530015K06Rik, SM-LmodApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-971