Learn More
Biomatik Corporation Lmp2 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11908
Encompass_Preferred
Lmp2 Polyclonal Antibody is a Epstein-Barr virus Polyclonal Antibody to LMP2. The Lmp2 Polyclonal Antibody has been validated for the following applications: ELISA. Host: Rabbit. Species: Epstein-Barr virus. Immunogen: Recombinant Epstein-Barr Virus Latent Membrane Protein 2 Protein (1-147Aa). Applications: ELISA. Target Synonyms: LMP2. Accession Number: P13285. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9228