Learn More
Medchemexpress LLC HY-111790 5mg Medchemexpress, LMP7-IN-1 CAS:2285330-15-4 Purity:>98%
SDP
Catalog No. 501968966
Encompass_Preferred
Medchemexpress, HY-111790 5mg LMP7-IN-1 CAS:2285330-15-4 LMP7-IN-1 is an inhibitor of immunoproteasome (LMP7), may used in the research of inflammatory and autoimmune diseases, neurodegenerative diseases, proliferative diseases and cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8966 Supplier Medchemexpress LLC Supplier No. HY1117905MG
Medchemexpress, HY-111790 5mg LMP7-IN-1 CAS:2285330-15-4 LMP7-IN-1 is an inhibitor of immunoproteasome (LMP7), may used in the research of inflammatory and autoimmune diseases, neurodegenerative diseases, proliferative diseases and cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.